6-Cyclopropylpyridine-3-carbonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR315871-1G OR315871-5G
APOSOR315871-1GEA 1700 DKK
APOSOR315871-1G APOSOR315871-5G
6-Cyclopropylpyridine-3-carbonitrile
6-Cyclopropylpyridine-3-carbonitrile
Formel: C₉H₈N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18255574
CAS-nummer: 1032527-28-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR