1-benzyl-3-methylbenzene 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Benzyl-3-methylbenzene
OR311219-1G
OR311219-250MG
APOSOR311219-1GEA
3520
DKK
APOSOR311219-1G
APOSOR311219-250MG
1-benzyl-3-methylbenzene 95%
1-benzyl-3-methylbenzene
Formel:
C₁₄H₁₄ Kogepunkt: 276 °C (760 mm Hg) Smeltepunkt: –34 °C Massefylde: 0,997 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09970508 CAS-nummer: 620-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...