2-(4-Pentylcyclohexyl)acetic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR310554-1G
OR310554-250MG
OR310554-5G
OR310554-2.5G
OR310554-10G
APOSOR310554-1GEA
3730
DKK
APOSOR310554-1G
APOSOR310554-250MG
APOSOR310554-5G
APOSOR310554-2.5G
APOSOR310554-10G
2-(4-Pentylcyclohexyl)acetic acid
2-(4-Pentylcyclohexyl)acetic acid
Formel:
C₁₃H₂₄O₂ Smeltepunkt: 60…62 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00995144 CAS-nummer: 74603-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...