2-{[(2,4-Dimethoxyphenyl)methyl]amino}acetonitrile hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR310331-1G OR310331-500MG
APOSOR310331-1GEA 7990 DKK
APOSOR310331-1G APOSOR310331-500MG
2-{[(2,4-Dimethoxyphenyl)methyl]amino}acetonitrile hydrochloride
2-{[(2,4-Dimethoxyphenyl)methyl]amino}acetonitrile hydrochloride
Formel: C₁₁H₁₄N₂O₂·ClH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26523309
CAS-nummer: 94459-09-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR