4-Chloro-2-trichloromethyl-quinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR307814-1G OR307814-5G
APOSOR307814-1GEA 1770 DKK
APOSOR307814-1G APOSOR307814-5G
4-Chloro-2-trichloromethyl-quinoline
4-Chloro-2-trichloromethyl-quinoline
Formel: C₁₀H₅Cl₄N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00272353
CAS-nummer: 35871-17-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR