3-Methyl-1-nitro-1H-pyrazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR307303-1G
OR307303-250MG
APOSOR307303-1GEA
763
DKK
APOSOR307303-1G
APOSOR307303-250MG
3-Methyl-1-nitro-1H-pyrazole
3-Methyl-1-nitro-1H-pyrazole
Formel:
C₄H₅N₃O₂ Molekylvægt: 127.101 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00466493 CAS-nummer: 31163-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...