6-Cyclopropylpicolinic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR306621-1G OR306621-250MG
APOSOR306621-1GEA 4310 DKK
APOSOR306621-1G APOSOR306621-250MG
6-Cyclopropylpicolinic acid
6-Cyclopropylpicolinic acid
Formel: C₉H₉NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18255566
CAS-nummer: 1256793-43-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR