4-(Chloromethyl)-1H-indazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR305069-250MG
OR305069-1G
APOSOR305069-250MGEA
915
DKK
APOSOR305069-250MG
APOSOR305069-1G
4-(Chloromethyl)-1H-indazole
4-(Chloromethyl)-1H-indazole
Formel:
C₈H₇ClN₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD10696785 CAS-nummer: 944898-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...