1-Methylazetidin-3-one hydrochloride
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR304316-1G
OR304316-5G
APOSOR304316-1GEA
2240
DKK
APOSOR304316-1G
APOSOR304316-5G
1-Methylazetidin-3-one hydrochloride
1-Methylazetidin-3-one hydrochloride
Formel:
C₄H₇NO·ClH Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD29060167 CAS-nummer: 1818847-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...