5-Ethyl-2-methylpiperidine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR30399-1G
OR30399-5G
APOSOR30399-1GEA
933
DKK
APOSOR30399-1G
APOSOR30399-5G
5-Ethyl-2-methylpiperidine
5-Ethyl-2-methylpiperidine
Formel:
C₈H₁₇N Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00023134 CAS-nummer: 104-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...