1-Methyl-5-nitropyrazole-4-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR3037-1G
OR3037-5G
APOSOR3037-1GEA
0
DKK
APOSOR3037-1G
APOSOR3037-5G
1-Methyl-5-nitropyrazole-4-carboxylic acid
1-Methyl-5-nitropyrazole-4-carboxylic acid
Formel:
C₅H₅N₃O₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00052540 CAS-nummer: 18213-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...