[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR303320-1G OR303320-250MG
APOSOR303320-1GEA 1150 DKK
APOSOR303320-1G APOSOR303320-250MG
[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester
[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester
Formel: C₁₁H₂₁BO₂Si
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13182130
CAS-nummer: 159087-46-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR