2-Chloroquinoxalin-6-amine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR300371-1G OR300371-250MG
APOSOR300371-1GEA 1430 DKK
APOSOR300371-1G APOSOR300371-250MG
2-Chloroquinoxalin-6-amine
2-Chloroquinoxalin-6-amine
Formel: C₈H₆ClN₃
Molekylvægt: 179.606 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16628196
CAS-nummer: 112928-27-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR