3-Bromo-5-chloroisonicotinonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR300370-1G OR300370-250MG
APOSOR300370-1GEA 3040 DKK
APOSOR300370-1G APOSOR300370-250MG
3-Bromo-5-chloroisonicotinonitrile
3-Bromo-5-chloroisonicotinonitrile
Formel: C₆H₂BrClN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22380416
CAS-nummer: 1335052-66-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR