3-Iodo-7-methoxy-1H-pyrrolo[2,3-c]pyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR300187-1G OR300187-250MG
APOSOR300187-1GEA 2510 DKK
APOSOR300187-1G APOSOR300187-250MG
3-Iodo-7-methoxy-1H-pyrrolo[2,3-c]pyridine
3-Iodo-7-methoxy-1H-pyrrolo[2,3-c]pyridine
Formel: C₈H₇IN₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12963282
CAS-nummer: 1190316-96-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR