1-Hydroxyisoquinoline-6-carbonitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Cyano-1-hydroxyisoquinoline, 6-Cyano-1-hydroxy-2-azanaphthalene
OR300072-250MG
OR300072-500MG
APOSOR300072-250MGEA
3080
DKK
APOSOR300072-250MG
APOSOR300072-500MG
1-Hydroxyisoquinoline-6-carbonitrile
1-Hydroxyisoquinoline-6-carbonitrile
Formel:
C₁₀H₆N₂O Molekylvægt: 170.167 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD22375647 CAS-nummer: 1184916-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...