1-Chloroisoquinoline-6-carbonitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-6-cyanoisoquinoline, 1-Chloro-6-cyano-2-azanaphthalene
OR300059-500MG
OR300059-250MG
APOSOR300059-500MGEA
3050
DKK
APOSOR300059-500MG
APOSOR300059-250MG
1-Chloroisoquinoline-6-carbonitrile
1-Chloroisoquinoline-6-carbonitrile
Formel:
C₁₀H₅ClN₂ Molekylvægt: 188.613 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD18250894 CAS-nummer: 1260664-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...