3-(Chloromethyl)-5-thien-3-yl-1,2,4-oxadiazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
OR29094-250MG
OR29094-100MG
APOSOR29094-250MGEA
1040
DKK
APOSOR29094-250MG
APOSOR29094-100MG
3-(Chloromethyl)-5-thien-3-yl-1,2,4-oxadiazole
3-(Chloromethyl)-5-thien-3-yl-1,2,4-oxadiazole
Formel:
C₇H₅ClN₂OS Smeltepunkt: 65/70 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00111629 CAS-nummer: 184970-24-9 UN nummer: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...