4-(Dimethylamino)chalcone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-[4-(Dimethylamino)phenyl]-1-phenylprop-2-en-1-one

OR27791-1G OR27791-250MG
APOSOR27791-1GEA 1860 DKK
APOSOR27791-1G APOSOR27791-250MG
4-(Dimethylamino)chalcone
4-(Dimethylamino)chalcone
Formel: C₁₇H₁₇NO
Molekylvægt: 251.323 g/mol
Smeltepunkt: 111…113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00168677
CAS-nummer: 1030-27-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR