7-methyl-1-naphthol 96%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR27682-250MG
OR27682-100MG
OR27682-25MG
APOSOR27682-250MGEA
8980
DKK
APOSOR27682-250MG
APOSOR27682-100MG
APOSOR27682-25MG
7-methyl-1-naphthol 96%
7-methyl-1-naphthol
Formel:
C₁₁H₁₀O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01314197 CAS-nummer: 6939-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...