6-Cyclobutoxypyridine-2-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR200163-1G OR200163-50MG OR200163-250MG OR200163-500MG
APOSOR200163-1GEA 6920 DKK
APOSOR200163-1G APOSOR200163-50MG APOSOR200163-250MG APOSOR200163-500MG
6-Cyclobutoxypyridine-2-carboxylic acid
6-Cyclobutoxypyridine-2-carboxylic acid
Formel: C₁₀H₁₁NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16697783
CAS-nummer: 1250298-25-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR