1-(Methylsulphonyl)piperidin-4-one

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(Methylsulphonyl)-4-oxopiperidine

OR16033-1G OR16033-250MG
APOSOR16033-1GEA 186 DKK
APOSOR16033-1G APOSOR16033-250MG
1-(Methylsulphonyl)piperidin-4-one
1-(Methylsulphonyl)piperidin-4-one
Formel: C₆H₁₁NO₃S
Molekylvægt: 177.221 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08061935
CAS-nummer: 218780-53-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR