4-[4-(3-Oxobutyl)phenoxy]benzonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR15758-10G OR15758-1G OR15758-5G
APOSOR15758-10GEA 4740 DKK
APOSOR15758-10G APOSOR15758-1G APOSOR15758-5G
4-[4-(3-Oxobutyl)phenoxy]benzonitrile
4-[4-(3-Oxobutyl)phenoxy]benzonitrile
Formel: C₁₇H₁₅NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11035754
CAS-nummer: 189120-04-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR