Propane-1-sulphonamide 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-(Sulphamoyl)propane
OR11393-5G
OR11393-10G
APOSOR11393-5GEA
741
DKK
APOSOR11393-5G
APOSOR11393-10G
Propane-1-sulphonamide 97%
Propane-1-sulphonamide
Formel:
C₃H₉NO₂S Smeltepunkt: 51…52 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD03550611 CAS-nummer: 24243-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...