N,N-Diisopropyl-4-pentylbenzamide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR111139-10G
OR111139-1G
OR111139-5G
APOSOR111139-10GEA
0
DKK
APOSOR111139-10G
APOSOR111139-1G
APOSOR111139-5G
N,N-Diisopropyl-4-pentylbenzamide
N,N-Diisopropyl-4-pentylbenzamide
Formel:
C₁₈H₂₉NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD28987629 CAS-nummer: 38630-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...