2,4-Dichloro-N,N-diisopropylbenzamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR111137-5G OR111137-10G OR111137-1G
APOSOR111137-5GEA 0 DKK
APOSOR111137-5G APOSOR111137-10G APOSOR111137-1G
2,4-Dichloro-N,N-diisopropylbenzamide
2,4-Dichloro-N,N-diisopropylbenzamide
Formel: C₁₃H₁₇Cl₂NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00439054
CAS-nummer: 212392-34-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR