3-[(4-Methylpiperazin-1-yl)methyl]aniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR110891-1G OR110891-5G
APOSOR110891-1GEA 0 DKK
APOSOR110891-1G APOSOR110891-5G
3-[(4-Methylpiperazin-1-yl)methyl]aniline
3-[(4-Methylpiperazin-1-yl)methyl]aniline
Formel: C₁₂H₁₉N₃
Molekylvægt: 205.299 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06804545
CAS-nummer: 198281-55-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR