4-[4-(tert-Butyl)phenoxy]benzyl alcohol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: {4-[4-(tert-Butyl)phenoxy]phenyl}methanol, 4-(tert-Butyl)-4'-(hydroxymethyl)diphenyl ether

OR110523-1G OR110523-5G
APOSOR110523-1GEA 2230 DKK
APOSOR110523-1G APOSOR110523-5G
4-[4-(tert-Butyl)phenoxy]benzyl alcohol
4-[4-(tert-Butyl)phenoxy]benzyl alcohol
Formel: C₁₇H₂₀O₂
Smeltepunkt: 50…52 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12469075
CAS-nummer: 1037141-25-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR