N-(7-Amino-1,1,3,3,6-pentamethyl-2,3-dihydro-1H-inden-5-yl)acetamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR110283-1G OR110283-5G
APOSOR110283-1GEA 2180 DKK
APOSOR110283-1G APOSOR110283-5G
N-(7-Amino-1,1,3,3,6-pentamethyl-2,3-dihydro-1H-inden-5-yl)acetamide
N-(7-Amino-1,1,3,3,6-pentamethyl-2,3-dihydro-1H-inden-5-yl)acetamide
Formel: C₁₆H₂₄N₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13191322
CAS-nummer: 1303890-35-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR