Methyl 4-amino-3-chloro-5-methylbenzoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR0597-1G OR0597-250MG OR0597-5G
APOSOR0597-1GEA 469 DKK
APOSOR0597-1G APOSOR0597-250MG APOSOR0597-5G
Methyl 4-amino-3-chloro-5-methylbenzoate
Methyl 4-amino-3-chloro-5-methylbenzoate
Formel: C₉H₁₀ClNO₂
Smeltepunkt: 80…82 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01313708
CAS-nummer: 202146-16-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR