α-Zearalenol Giberella zeae

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

BIZL0113-10MG BIZL0113-5MG BIZL0113-25MG
APOSBIZL0113-10MGEA 2130 DKK
APOSBIZL0113-10MG APOSBIZL0113-5MG APOSBIZL0113-25MG
α-Zearalenol Giberella zeae
α-Zearalenol
Formel: C₁₈H₂₄O₅
Molekylvægt: 320.38 g/mol
Smeltepunkt: 165…166 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD16657008
CAS-nummer: 36455-72-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR