Amino-PEG10-acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

BIPG1038-100MG BIPG1038-50MG
APOSBIPG1038-100MGEA 1030 DKK
APOSBIPG1038-100MG APOSBIPG1038-50MG
Amino-PEG10-acid
Amino-PEG10-acid
Formel: C₂₃H₄₇NO₁₂
Opbevaringstemperatur: Fryser
MDL Number: MFCD26127813
CAS-nummer: 2170987-85-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR