Dinatrium 4-nitrophenyl phosphat hexahydrat (pNPP)
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate
BIN0446-100G BIN0446-25G BIN0446-50G
APOSBIN0446-100GEA 1590 DKK
APOSBIN0446-100G APOSBIN0446-25G APOSBIN0446-50G
Dinatrium 4-nitrophenyl phosphat hexahydrat (pNPP)
Dinatrium 4-nitrophenyl phosphat hexahydrat (pNPP)
Formel: C₆H₄NNa₂O₆P·6H₂O
Molekylvægt: 371.144 g/mol
Smeltepunkt: >300 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00066288
CAS-nummer: 333338-18-4
EINECS: 224-246-5