Nivalenol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
BIN0113-1MG
BIN0113-5MG
APOSBIN0113-1MGEA
1370
DKK
APOSBIN0113-1MG
APOSBIN0113-5MG
Nivalenol
Nivalenol
Formel:
C₁₅H₂₀O₇ Smeltepunkt: 127…130 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00133673 CAS-nummer: 23282-20-4 UN nummer: 2811 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...