Muramic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (R)-2-(((2R,3R,4R,5R)-2-Amino-4,5,6-trihydroxy-1-oxohexan-3-yl)oxy)propanoic acid, 2-Amino-3-O-(1-carboxyethyl)-2-deoxy-D-glucose

BIMM1062-10MG BIMM1062-25MG
APOSBIMM1062-10MGEA 4800 DKK
APOSBIMM1062-10MG APOSBIMM1062-25MG
Muramic acid
Muramic acid
Formel: C₉H₁₇NO₇
Opbevaringstemperatur: Fryser
MDL Number: MFCD00213428
CAS-nummer: 1114-41-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR