Methyl 4-O-(α-D-galactopyranosyl)-α-D-galactopyranoside ≥98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BICL4255-10MG BICL4255-5MG
APOSBICL4255-10MGEA 5670 DKK
APOSBICL4255-10MG APOSBICL4255-5MG
Methyl 4-O-(α-D-galactopyranosyl)-α-D-galactopyranoside ≥98%
Methyl 4-O-(α-D-galactopyranosyl)-α-D-galactopyranoside
Formel: C₁₃H₂₄O₁₁
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD15145343
CAS-nummer: 67145-39-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR