Tolcapone 3-O-β-D-glucuronide ≥95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BICL4230-1MG BICL4230-2MG BICL4230-5MG
APOSBICL4230-1MGEA 2840 DKK
APOSBICL4230-1MG APOSBICL4230-2MG APOSBICL4230-5MG
Tolcapone 3-O-β-D-glucuronide ≥95%
Tolcapone 3-O-β-D-glucuronide
Formel: C₂₀H₁₉NO₁₁
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09952292
CAS-nummer: 204853-33-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR