4-Nitrophenyl-β-D-mannopyranoside
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
BIB6133-100MG
BIB6133-1G
BIB6133-250MG
APOSBIB6133-100MGEA
1480
DKK
APOSBIB6133-100MG
APOSBIB6133-1G
APOSBIB6133-250MG
4-Nitrophenyl-β-D-mannopyranoside
4-Nitrophenyl-β-D-mannopyranoside
Formel:
C₁₂H₁₅NO₈ Molekylvægt: 301.249 g/mol Smeltepunkt: 207 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00067650 CAS-nummer: 35599-02-1 EINECS: 252-633-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...