6-Methoxyindole-3-acetic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
BIB6101-1G
BIB6101-500MG
BIB6101-2.5G
APOSBIB6101-1GEA
556
DKK
APOSBIB6101-1G
APOSBIB6101-500MG
APOSBIB6101-2.5G
6-Methoxyindole-3-acetic acid
6-Methoxyindole-3-acetic acid
Formel:
C₁₁H₁₁NO₃ Smeltepunkt: 163…165 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01548400 CAS-nummer: 103986-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...