Chemo gowns, Tyvek® IsoClean®, model IC703S 00, non sterile
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IC703SWH00/S IC703SWH00/XS IC703SWH00/M IC703SWH00/L IC703SWH00/XL IC703SWH00/2XL IC703SWH00/3XL
DUPOIC703SWH00/SEA 2530 DKK
DUPOIC703SWH00/S DUPOIC703SWH00/XS DUPOIC703SWH00/M DUPOIC703SWH00/L DUPOIC703SWH00/XL DUPOIC703SWH00/2XL DUPOIC703SWH00/3XL
Chemo gowns, Tyvek® IsoClean®, model IC703S 00, non sterile
Engangsbeklædning
Collared gowns with hook and loop closure in the neck. Increased skirt length ensures protection against frontal exposure. Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams, knit cuffs
  • Openness in the back closure increases wearer comfort
  • Not clean-processed and not sterilised
  • Bound ties at waist originating from elasticated sides
  • Ideal for handling cytostatics and various laboratory activities
  • Made of high density polyethylene using a patented flash spinning process
Tyvek® IsoClean® provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash.
Tyvek® IsoClean® (option codes 0B, 00 and BH) garments and accessories are neither clean-processed nor gamma-irradiated but manufactured in a controlled environment.
Garments and accessories made of Tyvek® IsoClean® are typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Certificeringer: (EU) 2016/425; Cat III, Type PB [6-B]; EN 14126. Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
Emballering: Bulk packaged in double transparent poly liners. 30 gowns per case.