Graduated wide neck square bottles, PE, with screw cap and undercap
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
608 609 610 611 612 613 614
216-2565EA 4290 DKK
216-2565 216-2566 216-2567 216-2568 216-2569 216-2570 216-2508
Graduated wide neck square bottles, PE, with screw cap and undercap
Flasker Plastflasker
These high quality graduated, wide neck square bottles are made of PE (main body and under cap) and supplied with a PP cap.
  • Suitable for foodstuff contact according to EC Regulation 10/2011
  • Translucent
  • Tagging points on shoulder and cap ensures tamper-evidence by means of tags and seals
Graduated space saving bottles, ideal for long-term storage of solid or liquid samples. Sloping shoulders reduce tendency for residue deposits. Moulded-in graduations.
Certificeringer: Meets EC Regulation 10/2011. Migration Test Compliance Declaration available.