VWR®, Measuring Flasks With Single Certificate
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-6701EA 279 DKK
612-6701 612-6702 612-6703 612-6704 612-6705 612-6706 612-6707 612-6708 612-6709 612-6710 612-6711
VWR®, Measuring Flasks With Single Certificate
Kolber Volumetriske kolber
Borosilicate 3.3 glass, with PP stopper. Single certified class A measuring flasks with imprinted serial number and individual work certificate.
  • High chemical and thermal resistance
  • Delivered with white PP stopper
  • Inscriptions and ring mark in high contrasting blue enamel
  • Calibrated to contain (TC, In)
  • Imprinted serial number and single certificate included
Made from true bore glass tubing to guarantee exact measurement.
Certificeringer: DIN EN ISO 1042. Conformity certified with DE-M mark.