VWR® Bulb Pipettes, One Mark, Class AS, with Single Certificate
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-6712EA 246 DKK
612-6712 612-6713 612-6714 612-6715 612-6716 612-6717 612-6718 612-6719 612-6720 612-6721
VWR® Bulb Pipettes, One Mark, Class AS, with Single Certificate
Pipetter Kolbepipetter
Borosilicate 9.1 glass. Single certified class AS bulb pipettes with one mark, imprinted serial number and individual work certificate.
  • Calibrated to deliver (TD, Ex)
  • Inscriptions and ring mark in high contrasting amber
  • Imprinted serial number, single certificate included
Made from true bore glass tubing to guarantee exact measurement at each measuring point. Colour coded and conformity certified with DE-M mark.
Certificeringer: DIN EN ISO 648