VWR® VINYL HYBRID, Gloves, Vinyl
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112-6072EA 115 DKK
112-6072 112-6073
VWR® VINYL HYBRID, Gloves, Vinyl
Handsker
A glove lower in price than nitrile and higher in quality than vinyl.
  • Non sterile
  • Powder-free
  • Particularly skin friendly for users with latex allergy
  • Rolled cuffs minimise the risk of tearing
These innovative vinyl gloves are made from a revolutionary new formula of vinyl-based hybrid material. The formulation combines vinyl and nitrile properties in one glove by using the best of each material and providing outstanding value. These gloves offer enhanced tactile sensitivity and good barrier protection for a wide range of applications, such as food handling, catering and kitchen work, lab applications, and other tasks with complex risks.
Certificeringer: EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS; Cat. III
Food contact approved.
Advarsel: * Thickness is double measured, minimum values.