Safety spectacles, 3700 series, SecureFit™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SF3707XSGAFBLU SF3702SGAF-BLU SF3701ASP-BLU SF3701SGAF-BLU SF3701SGAF-GRN SF3701SGAF-ORG SF3701XSGAFBLU SF3707SGAF-BLU SF3702XSGAFBLU SF3703SGAF-GRN
111-2529EA 280 DKK
111-2529 111-2525 111-2520 111-2521 111-2522 111-2523 111-2524 111-2528 111-2526 111-2527
Safety spectacles, 3700 series, SecureFit™
Briller
SecureFit™ 3700 series are safety over-spectacles designed for use with prescription glasses. These lightweight over-spectacles are designed to fit over the users existing glasses to provide personalised protection.
  • Adjustable ratchet temples help to ensure a secure fit
  • Pressure Diffusion Temples (PDT) reduce pressure over the ear for a secure and comfortable fit
  • Polycarbonate lenses absorb 99,9% of UVA and UVB rays
  • Anti-scratch and anti-fog coatings provide increased durability and improved vision
  • Similar to typical sunglasses, grey lenses are popular in outdoor daytime work environments
For safety spectacles that fit over prescription glasses and feature adjustable ratchet temples, choose the SecureFit™ 3700 series. The adjustable ratchet temples provide angle adjustment points on the temple to ensure a secure fit. They also feature Pressure Diffusion Temple (PDT) technology and a soft touch nosepiece to help provide all-day comfort. The polycarbonate lens is available with Scotchgard™ anti-fog and anti-scratch coatings for increased durability and improved vision. Spectacles are available in three temple arm colours and four lens options. Also available with a contoured brow guard.
Certificeringer: Meets the requirements of EN 166-2001 standard as an impact rated spectacle.