Leucoberbelin Blue I 65% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: LBB

432199-100MG 432199-500MG
432199-100MGEA 708 DKK
432199-100MG 432199-500MG
Leucoberbelin Blue I 65% (dye content), Sigma-Aldrich®
Leucoberbelin Blue I

Leucoberbelin blue I has been used as a quencher in time-resolved fluorescence QRET technique for receptor-ligand interaction screening and to detect oxidized manganese species in Mn(II) (manganese) oxidation assays.


Leucoberbelin Blue I is a colourimetric reagent redox dye. It is a water-soluble leuco base also abbreviated as LBB. Upon oxidation, LBB forms a coloured complex which is stoichiometrically related to the oxidation state of the reagent.

Formel: C₂₃H₂₆N₂O₃S
Molekylvægt: 410.53 g/mol
Smeltepunkt: >300 °C (lit.)
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00217522
CAS-nummer: 52748-86-4
UN nummer: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d85579b5f44f1e6bea105de8cbbd761aa41190d&vid=5d85579b5f44f1e6bea105de8cbbd761aa41190d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light blue powder
Infrared spectrum Conforms to structure and standard as Illustrated on page 4132C of edition I, Volume 3 of 'the aldrich library of FT-IR Spectra'.
Elemental analysis Carbon 42.4% (minimum), Nitrogen 4.3% (minimum)
Thin-layer Consistent with control


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR