Ethyl viologen dibromide 99%, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1'-Diethyl-[4,4'-bipyridine]-1,1'-diium bromide, 1,1'-Diethyl-4,4'-bipyridinium dibromide

384097-1G 384097-5G
384097-1GEA 434 DKK
384097-1G 384097-5G
Ethyl viologen dibromide 99%, Sigma-Aldrich®
Ethyl viologen dibromide

Ethyl viologen dibromide (1,1-diethyl- 4,4-bipyridinium dibromide) has been used as an internal standard in analysis of paraquat and diquat in the serum using high-performance liquid chromatography.

Formel: C₁₄H₁₈Br₂N₂
Molekylvægt: 374.12 g/mol
Smeltepunkt: 278 °C (dec., lit.)
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00134590
CAS-nummer: 53721-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12db0cafe6531014c0adbdb51d9159e91c33d0d3&vid=12db0cafe6531014c0adbdb51d9159e91c33d0d3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Yellow powder, crystals, crystalline powder and/or chunks
Infrared spectrum Conforms to structure
Titration 98.5%-101.5% (with silver nitrate)
High pressure liquid 98.5% (minimum)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR