8-Hydroxyjulolidine 96%, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPYSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol
8-Hydroxyjulolidine has been used in the electrochemical switching fluorescence study.
The carboxaldehyde derivative of 8-hydroxyjulolidine is useful as a chromophore in fluorescence chemosensor and chemosensors.
Formel:
C₁₂H₁₅NO Opbevaringstemperatur: Room Temperature |
MDL Number:
MFCD00006918 CAS-nummer: 41175-50-2 EINECS: 255-247-9 |
|
Specifikationer Testresultater
Appearance (Colour) | Off White to Dark Beige and Faint Orange to Orange and Pink |
Appearance (Form) | Powder or Crystals |
Infrared spectrum | Conforms to Structure |
Carbon | 72.7 - 79.6% |
Nitrogen | 7.1 - 7.7% |
Purity (HPLC) | ≥95.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...