Hoods and hood assemblies S-400 series PAPR, Versaflo™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S433L S433S
111-0850EA 1580 DKK
111-0850 111-0851
Hoods and hood assemblies S-400 series PAPR, Versaflo™
Åndedrætsværn Åndedrætsbeskyttelsessystemer
Versaflo™ S-series hoods and headcovers feature adjustable suspensions (most models) and soft comfort pads that provide excellent fit, stability and tracking.
  • Can be used in dozens of different powered air turbo and supplied air respirator combinations
  • Direct a comfortable stream of air into the head tops
  • Features the innovative 3M™ Quick Release Swivel (QRS) connection
  • Closely profiles the wearer’s head for a more streamlined, less bulky appearance
Suspension hoods and head covers include wide face seals with soft elastic for improved comfort and reduced airflow noise. Reusable suspension allows wearers to modify airflow distribution inside the hood to suit personal comfort needs. Offering eye, face, hair, neck and shoulder protection, S-400 series hoods are intrinsically safe in environments requiring high mobility. Hoods feature a two-layer polypropylene spunbonded/laminated film fabric construction, with PETG visor. They provide protection against gas, vapour, particulates, liquid splash, dust and low energy flying particles.
Certificeringer: Meets EN 12941 TH3, EN 166:2:F:3, EN 14594 3A.