Thiacloprid
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
464390010
464392500
ACRO464390010EA
1450
DKK
ACRO464390010
ACRO464392500
Thiacloprid
Thiacloprid
- Bioactive
Formel:
C₁₀H₉ClN₄S Molekylvægt: 252.72 g/mol Smeltepunkt: 133…136 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD02101042 CAS-nummer: 111988-49-9 UN nummer: 2811 ADR: 6.1 III |
Specifikationer Testresultater
Appearance | Powder |
Infrared spectrum | Conforms |
Melting point | 133 to 138 °C |
Water | ≤1 % |
HPLC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...